CAS RN 1873687-86-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N,N-Diethyl-2-fluoro-6-nitroaniline, 97%, Thermo Scientific™
CAS: 1873687-86-5 Synonym: Diethyl-(2-fluoro-6-nitrophenyl)amine