accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNNHOOH

CAS RN 187400-85-7

IUPAC Name: 4-{5-[4-phenyl-8-(propan-2-yl)quinolin-2-yl]-1H-pyrrol-2-yl}benzoic acid
Synonyms: 4-[5-(8-isopropyl-4-phenylquinolin-2-yl)-1H-pyrrol-2-yl]benzoic acid
Molecular Weight (g/mol): 432.52
Molecular Formula: C29H24N2O2
InChi Key: LSGNKLDHMQVTEK-UHFFFAOYSA-N
SMILES: CC(C)C1=C2N=C(C=C(C3=CC=CC=C3)C2=CC=C1)C1=CC=C(N1)C1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results