accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 187665-65-2

IUPAC Name: 1-{2-[4-(4-carbamoylphenyl)piperazin-1-yl]ethyl}-N-methyl-3,4-dihydro-1H-2-benzopyran-6-carboxamide
Synonyms: 1-{2-[4-(4-carbamoylphenyl)piperazin-1-yl]ethyl}-N-methyl-3,4-dihydro-1H-2-benzopyran-6-carboxamide
Molecular Weight (g/mol): 422.53
Molecular Formula: C24H30N4O3
InChi Key: PNTVCCRNJOGKGA-UHFFFAOYNA-N
SMILES: CNC(=O)C1=CC2=C(C=C1)C(CCN1CCN(CC1)C1=CC=C(C=C1)C(N)=O)OCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)