CAS RN 18791-99-6
IUPAC Name:
4-bromothiophene-2-carbonitrile
Synonyms:
4-bromothiophene-2-carbonitrile
Molecular Weight (g/mol):
188.04
Molecular Formula:
C5H2BrNS
InChi Key:
DJYVXBJLHPKUKS-UHFFFAOYSA-N
SMILES:
BrC1=CSC(=C1)C#N