accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18824-63-0

IUPAC Name: 1,1-dimethoxynonane
Synonyms: 1,1-dimethoxynonane
Molecular Weight (g/mol): 188.31
Molecular Formula: C11H24O2
InChi Key: LYLVOCPDQAOQKL-UHFFFAOYSA-N
SMILES: CCCCCCCCC(OC)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results