Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonanal Dimethyl Acetal 97.0+%, TCI America™
SDP

Catalog No. N09155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09155G 5 g
N091525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09155G Supplier TCI America Supplier No. N09155G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18824-63-0
Molecular Formula C11H24O2
Molecular Weight (g/mol) 188.311
MDL Number MFCD00036646
InChI Key LYLVOCPDQAOQKL-UHFFFAOYSA-N
Synonym 1,1-Dimethoxynonane, Nonyl Aldehyde Dimethyl Acetal, Pelargonaldehyde Dimethyl Acetal
PubChem CID 87813
IUPAC Name 1,1-dimethoxynonane
SMILES CCCCCCCCC(OC)OC

Specifications

Color Colorless
Boiling Point 88°C
Quantity 5 g
Formula Weight 188.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonanal Dimethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.