accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHNONCl

CAS RN 188425-85-6

IUPAC Name: 2-chloro-N-{4'-chloro-[1,1'-biphenyl]-2-yl}pyridine-3-carboxamide
Synonyms: boscalid
Molecular Weight (g/mol): 343.21
Molecular Formula: C18H12Cl2N2O
InChi Key: WYEMLYFITZORAB-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=CC=CC=C1NC(=O)C1=CC=CN=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results