Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boscalid 98.0+%, TCI America™
SDP

Catalog No. B40381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40381G 1 g
B4038100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40381G Supplier TCI America Supplier No. B40381G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 188425-85-6
Molecular Formula C18H12Cl2N2O
Molecular Weight (g/mol) 343.207
MDL Number MFCD06795150
InChI Key WYEMLYFITZORAB-UHFFFAOYSA-N
Synonym 2-Chloro-N-(4′C-chlorobiphenyl-2-yl)nicotinamide
PubChem CID 213013
ChEBI CHEBI:81822
IUPAC Name 2-chloro-N-[2-(4-chlorophenyl)phenyl]pyridine-3-carboxamide
SMILES C1=CC=C(C(=C1)C2=CC=C(C=C2)Cl)NC(=O)C3=C(N=CC=C3)Cl

Specifications

Melting Point 149°C
Color White
Quantity 1 g
UN Number 3077
Formula Weight 343.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Boscalid
RTECSNumber US4587550
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.