accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHOHOO

CAS RN 1885-13-8

IUPAC Name: 4-methoxybenzene-1,2-dicarboxylic acid
Synonyms: 4-methoxybenzene-1,2-dicarboxylic acid
Molecular Weight (g/mol): 196.16
Molecular Formula: C9H8O5
InChi Key: JKZSIEDAEHZAHQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(O)=O)C(=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results