Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphthalic Acid 98.0+%, TCI America™
SDP

Catalog No. M14325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14325G 5 g
M14321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14325G Supplier TCI America Supplier No. M14325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1885-13-8
Molecular Formula C9H8O5
Molecular Weight (g/mol) 196.16
MDL Number MFCD00094739
InChI Key JKZSIEDAEHZAHQ-UHFFFAOYSA-N
PubChem CID 74665
IUPAC Name 4-methoxybenzene-1,2-dicarboxylic acid
SMILES COC1=CC=C(C(O)=O)C(=C1)C(O)=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 196.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphthalic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.