accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18851-33-7

IUPAC Name: 1,10-phenanthroline hydrate hydrochloride
Synonyms: phen hydrate hydrochloride
Molecular Weight (g/mol): 234.68
Molecular Formula: C12H11ClN2O
InChi Key: NDLHUHRGAIHALB-UHFFFAOYSA-N
SMILES: O.Cl.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Molecular Weight (g/mol) 
  • (4)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (5)
  • (3)
  • (2)
Quantity 
  • (3)
  • (2)
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (4)
  • (6)
  • (4)
  • (5)
  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (3)