accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHClONH3CCH3

CAS RN 188589-66-4

IUPAC Name: N-(2-chlorophenyl)-1,4a,6a-trimethyl-2-oxo-1H,2H,4aH,4bH,5H,6H,6aH,7H,8H,9H,9aH,9bH,10H,11H,11aH-indeno[5,4-f]quinoline-7-carboxamide
Synonyms: N-(2-chlorophenyl)-1,4a,6a-trimethyl-2-oxo-4bH,5H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-indeno[5,4-f]quinoline-7-carboxamide
Molecular Weight (g/mol): 441.01
Molecular Formula: C26H33ClN2O2
InChi Key: CTVXDPDUOKQBKZ-UHFFFAOYNA-N
SMILES: CN1C2CCC3C4CCC(C(=O)NC5=CC=CC=C5Cl)C4(C)CCC3C2(C)C=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results