accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOHOOHOOO

CAS RN 1887-02-1

IUPAC Name: disodium 4,6-dihydroxy-2,5-dioxocyclohexa-1(6),3-diene-1,3-bis(olate)
Synonyms: disodium 4,6-dihydroxy-2,5-dioxocyclohexa-1(6),3-diene-1,3-bis(olate)
Molecular Weight (g/mol): 216.06
Molecular Formula: C6H2Na2O6
InChi Key: SOSMJTGSYNGZAV-UHFFFAOYSA-L
SMILES: [Na+].[Na+].OC1=C([O-])C(=O)C([O-])=C(O)C1=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results