CAS RN 1888-57-9
IUPAC Name:
2,5-dimethylhexan-3-one
Synonyms:
3-hexanone, 2,5-dimethyl-
Molecular Weight (g/mol):
128.22
Molecular Formula:
C8H16O
InChi Key:
TUIWMHDSXJWXOH-UHFFFAOYSA-N
SMILES:
CC(C)CC(=O)C(C)C
1
–
1
of
1
results
1
2,5-Dimethyl-3-hexanone 96.0+%, TCI America™
CAS: 1888-57-9 Molecular Formula: C8H16O Molecular Weight (g/mol): 128.215 MDL Number: MFCD00048317 InChI Key: TUIWMHDSXJWXOH-UHFFFAOYSA-N Synonym: Isobutyl Isopropyl Ketone PubChem CID: 15901 IUPAC Name: 2,5-dimethylhexan-3-one SMILES: CC(C)CC(=O)C(C)C