accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18881-17-9

IUPAC Name: (3S)-3-(hydroxymethyl)-1,2,3,4-tetrahydroisoquinolin-2-ium
Synonyms: (3S)-3-(hydroxymethyl)-1,2,3,4-tetrahydroisoquinolin-2-ium
Molecular Weight (g/mol): 164.23
Molecular Formula: C10H14NO
InChi Key: ZSKDXMLMMQFHGW-JTQLQIEISA-O
SMILES: OC[C@@H]1CC2=CC=CC=C2C[NH2+]1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)