Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol 97.0+%, TCI America™
SDP

Catalog No. T17551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17551G Supplier TCI America Supplier No. T17551G

Chemical Identifiers

CAS 18881-17-9
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD01631316
InChI Key ZSKDXMLMMQFHGW-JTQLQIEISA-N
Synonym s-1,2,3,4-tetrahydroisoquinoline-3-methanol, 3s-1,2,3,4-tetrahydroisoquinolin-3-ylmethanol, s-1,2,3,4-tetrahydroisoquinolin-3-yl methanol, 3s-1,2,3,4-tetrahydroisoquinolin-3-yl methanol, chembl61489, s-1,2,3,4-tetrahydroisoquinolin-3-yl-methanol, s---1,2,3,4-tetrahydro-3-isoquinolinemethanol, s-3-hydroxymethyl-1,2,3,4-tetrahydroisoquinoline, 3-isoquinolinemethanol, 1,2,3,4-tetrahydro-, s, s-1,2,3,4-tetrahydroisoquinolylmethan-3-ol
PubChem CID 776757
IUPAC Name [(3S)-1,2,3,4-tetrahydroisoquinolin-3-yl]methanol
SMILES C1C(NCC2=CC=CC=C21)CO

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 1 g
Formula Weight 163.22
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.