accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNFFFCl

CAS RN 188817-13-2

IUPAC Name: 5-(4-chlorophenyl)-1-(4-methoxyphenyl)-3-(trifluoromethyl)-1H-pyrazole
Synonyms: 5-(4-chlorophenyl)-1-(4-methoxyphenyl)-3-(trifluoromethyl)pyrazole
Molecular Weight (g/mol): 352.74
Molecular Formula: C17H12ClF3N2O
InChi Key: PQUGCKBLVKJMNT-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N1N=C(C=C1C1=CC=C(Cl)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results