accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 188923-01-5

IUPAC Name: ({5-[(2-{1-[3,5-bis(trifluoromethyl)phenyl]ethoxy}-3-(4-fluorophenyl)morpholin-4-yl)methyl]-2H-1,2,3-triazol-4-yl}methyl)dimethylamine hydrochloride
Synonyms: ({5-[(2-{1-[3,5-bis(trifluoromethyl)phenyl]ethoxy}-3-(4-fluorophenyl)morpholin-4-yl)methyl]-2H-1,2,3-triazol-4-yl}methyl)dimethylamine hydrochloride
Molecular Weight (g/mol): 611.99
Molecular Formula: C26H29ClF7N5O2
InChi Key: VZBKOBSSEVXFNF-UHFFFAOYNA-N
SMILES: Cl.CC(OC1OCCN(CC2=NNN=C2CN(C)C)C1C1=CC=C(F)C=C1)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)