accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 188970-92-5

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-{[(prop-2-en-1-yloxy)carbonyl]amino}propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-{[(prop-2-en-1-yloxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 410.43
Molecular Formula: C22H22N2O6
InChi Key: MPVGCCAXXFLGIU-UHFFFAOYNA-N
SMILES: OC(=O)C(CNC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)