accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 188975-30-6

IUPAC Name: 3,6-dihydro-2H-pyran-4-yl trifluoromethanesulfonate
Synonyms: 3,6-dihydro-2H-pyran-4-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 232.17
Molecular Formula: C6H7F3O4S
InChi Key: QXCAHEXKUMUTRF-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results