Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Dihydro-2H-pyran-4-yl trifluoromethanesulfonate, 95%, Thermo Scientific™

Catalog No. MO08368DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO08368DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO08368DA Supplier Thermo Scientific Chemicals Supplier No. MO08368DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 188975-30-6
Molecular Formula C6H7F3O4S
Molecular Weight (g/mol) 232.173
InChI Key QXCAHEXKUMUTRF-UHFFFAOYSA-N
Synonym 3,6-dihydro-2h-pyran-4-yl trifluoro-methanesulfonate, 5,6-dihydro-4-trifluoromethylsulfonyloxy-2h-pyran, 3,6-dihydro-2h-pyran-4-yltrifluoromethanesulfonate, 2,3-dihydro-4-trifluoromethylsulfonyloxy-6h-pyran, 2h-5,6-dihydropyran-4-yl trifluoromethyl sulfonate, 4-trifluoromethylsulfonyloxy-5,6-dihydro-2h-pyran, trifluoro-methanesulfonic acid 3,6-dihydro-2h-pyran-4-yl ester, trifluoromethanesulfonic acid 3,6-dihydro-2h-pyran-4-yl ester, methanesulfonic acid, 1,1,1-trifluoro-, 3,6-dihydro-2h-pyran-4-yl ester
PubChem CID 11356703
IUPAC Name 3,6-dihydro-2H-pyran-4-yl trifluoromethanesulfonate
SMILES C1COCC=C1OS(=O)(=O)C(F)(F)F

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 232.18
Chemical Name or Material 3, 6-Dihydro-2H-pyran-4-yl trifluoromethanesulfonate
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.