accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHNOClClHCl

CAS RN 189061-11-8

IUPAC Name: N-{4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butyl}-9H-fluorene-2-carboxamide hydrochloride
Synonyms: N-{4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butyl}-9H-fluorene-2-carboxamide hydrochloride
Molecular Weight (g/mol): 530.92
Molecular Formula: C28H30Cl3N3O
InChi Key: PFIWYJNBKGCVFM-UHFFFAOYSA-N
SMILES: Cl.ClC1=CC=CC(N2CCN(CCCCNC(=O)C3=CC4=C(C=C3)C3=CC=CC=C3C4)CC2)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)