accessibility menu, dialog, popup

UserName

Viewing profile as user:
OPOBrClO

CAS RN 189099-56-7

IUPAC Name: diphenyl [(4-bromophenyl)(chloro)methyl]phosphonate
Synonyms: diphenyl (4-bromophenyl)(chloro)methylphosphonate
Molecular Weight (g/mol): 437.65
Molecular Formula: C19H15BrClO3P
InChi Key: ZSYMFYMYJVSPDH-UHFFFAOYNA-N
SMILES: ClC(C1=CC=C(Br)C=C1)P(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results