accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOOHOOHOHOHOHHOOHHOHO

CAS RN 18916-17-1

IUPAC Name: 1-(4-{[4,5-dihydroxy-6-(hydroxymethyl)-3-[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]oxan-2-yl]oxy}-2,6-dihydroxyphenyl)-3-(4-hydroxyphenyl)propan-1-one
Synonyms: 1-(4-{[4,5-dihydroxy-6-(hydroxymethyl)-3-[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]oxan-2-yl]oxy}-2,6-dihydroxyphenyl)-3-(4-hydroxyphenyl)propan-1-one
Molecular Weight (g/mol): 582.56
Molecular Formula: C27H34O14
InChi Key: CWBZAESOUBENAP-UHFFFAOYNA-N
SMILES: CC1OC(OC2C(O)C(O)C(CO)OC2OC2=CC(O)=C(C(=O)CCC3=CC=C(O)C=C3)C(O)=C2)C(O)C(O)C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results