accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1892-29-1

IUPAC Name: 2-[(2-hydroxyethyl)disulfanyl]ethan-1-ol
Synonyms: 2-hydroxyethyl disulfide heds compound 2-hydroxyethanedisulfide usaf TH-9 2,2'-disulfanediyldiethanol
Molecular Weight (g/mol): 154.24
Molecular Formula: C4H10O2S2
InChi Key: KYNFOMQIXZUKRK-UHFFFAOYSA-N
SMILES: OCCSSCCO
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (6)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)