accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3N

CAS RN 18936-17-9

IUPAC Name: 2-methylbutanenitrile
Synonyms: 2-methylbutyronitrile 2-methylbutanone cyanohydrin s-butyl cyanide
Molecular Weight (g/mol): 83.13
Molecular Formula: C5H9N
InChi Key: RCEJCSULJQNRQQ-UHFFFAOYNA-N
SMILES: CCC(C)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results