CAS RN 189750-98-9
IUPAC Name:
4-ethoxy-2,3-difluoro-4'-(4-propylcyclohexyl)-1,1'-biphenyl
Synonyms:
4-ethoxy-2,3-difluoro-4'-(4-propylcyclohexyl)-1,1'-biphenyl
Molecular Weight (g/mol):
358.47
Molecular Formula:
C23H28F2O
InChi Key:
IBFAIOMGVHPWRQ-UHFFFAOYSA-N
SMILES:
CCCC1CCC(CC1)C1=CC=C(C=C1)C1=C(F)C(F)=C(OCC)C=C1
1
–
1
of
1
results
1
4-Ethoxy-2,3-difluoro-4'-(trans-4-propylcyclohexyl)biphenyl 98.0+%, TCI America™
CAS: 189750-98-9 Molecular Formula: C23H28F2O Molecular Weight (g/mol): 358.473 MDL Number: MFCD18451991 InChI Key: IBFAIOMGVHPWRQ-UHFFFAOYSA-N PubChem CID: 18681376 IUPAC Name: 1-ethoxy-2,3-difluoro-4-[4-(4-propylcyclohexyl)phenyl]benzene SMILES: CCCC1CCC(CC1)C2=CC=C(C=C2)C3=C(C(=C(C=C3)OCC)F)F