accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18982-54-2

IUPAC Name: (2-bromophenyl)methanol
Synonyms: benzenemethanol, 2-bromo-
Molecular Weight (g/mol): 187.04
Molecular Formula: C7H7BrO
InChi Key: IOWGHQGLUMEZKG-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1Br
Molecular Weight (g/mol) 
  • (1)
  • (7)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (5)
  • (1)
  • (7)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)