Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzyl alcohol, 98%, Thermo Scientific™

Catalog No. AC180100500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC180100500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC180100500 Supplier Thermo Scientific Chemicals Supplier No. 180100500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18982-54-2
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00004600
InChI Key IOWGHQGLUMEZKG-UHFFFAOYSA-N
Synonym 2-bromobenzyl alcohol, 2-bromophenyl methanol, o-bromobenzyl alcohol, benzenemethanol, 2-bromo, o-bromo-benzyl alcohol, 2-bromobenzylalcohol, 2-bromophenyl methan-1-ol, o-bromobenzylalcohol, 2-bromo-benzylalcohol, pubchem3746
PubChem CID 72850
IUPAC Name (2-bromophenyl)methanol
SMILES OCC1=CC=CC=C1Br

Specifications

Melting Point 78.0°C to 82.0°C
Linear Formula BrC6H4CH2OH
Quantity 50 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most organic solvents
Formula Weight 187.04
Chemical Name or Material 2-Bromobenzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.