accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CCCH3NH3CCClPdClPd

CAS RN 18987-59-2

IUPAC Name: bis(2-[(dimethylamino)methyl]benzen-1-ide); bis(chloropalladiumylium)
Synonyms: bis(2-[(dimethylamino)methyl]benzen-1-ide); bis(chloropalladiumylium)
Molecular Weight (g/mol): 552.14
Molecular Formula: C18H24Cl2N2Pd2
InChi Key: RYOPVFUGUFDYKP-UHFFFAOYSA-L
SMILES: Cl[Pd+].Cl[Pd+].CN(C)CC1=CC=CC=[C-]1.CN(C)CC1=CC=CC=[C-]1
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)