accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOBrOOH3C

CAS RN 1906-95-2

IUPAC Name: 1,3-diethyl 2-(5-bromopentyl)propanedioate
Synonyms: 1,3-diethyl 2-(5-bromopentyl)propanedioate
Molecular Weight (g/mol): 309.2
Molecular Formula: C12H21BrO4
InChi Key: JZCAZGWJCVMTTQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CCCCCBr)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results