accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 191282-48-1

IUPAC Name: 4-methylidene-2-octyl-5-oxooxolane-3-carboxylic acid
Synonyms: 4-methylidene-2-octyl-5-oxooxolane-3-carboxylic acid
Molecular Weight (g/mol): 254.33
Molecular Formula: C14H22O4
InChi Key: VCWLZDVWHQVAJU-UHFFFAOYNA-N
SMILES: CCCCCCCCC1OC(=O)C(=C)C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results