accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 191471-52-0

IUPAC Name: 4-amino-2-oxabicyclo[3.1.0]hexane-4,6-dicarboxylic acid
Synonyms: 4-amino-2-oxabicyclo[3.1.0]hexane-4,6-dicarboxylic acid
Molecular Weight (g/mol): 187.15
Molecular Formula: C7H9NO5
InChi Key: YASVRZWVUGJELU-UHFFFAOYNA-N
SMILES: NC1(COC2C(C12)C(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)