accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ONOHNOO

CAS RN 19171-19-8

IUPAC Name: 4-amino-2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: pomalidomide
Molecular Weight (g/mol): 273.25
Molecular Formula: C13H11N3O4
InChi Key: UVSMNLNDYGZFPF-UHFFFAOYNA-N
SMILES: NC1=CC=CC2=C1C(=O)N(C1CCC(=O)NC1=O)C2=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)