accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3CH3N

CAS RN 19219-98-8

IUPAC Name: 2,5,6-trimethyl-1,3-benzoxazole
Synonyms: 2,5,6-trimethyl-1,3-benzoxazole
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: PDVDINPEWGXOHX-UHFFFAOYSA-N
SMILES: CC1=NC2=CC(C)=C(C)C=C2O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results