CAS RN 192927-92-7
IUPAC Name:
1-{2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl}-3,3-dimethyl-2,3-dihydro-1H-indol-2-one hydrochloride
Synonyms:
1-{2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl}-3,3-dimethylindol-2-one hydrochloride
Molecular Weight (g/mol):
430.95
Molecular Formula:
C24H28ClFN2O2
InChi Key:
BOCLFQZPFYNVFD-UHFFFAOYSA-N
SMILES:
Cl.CC1(C)C(=O)N(CCN2CCC(CC2)C(=O)C2=CC=C(F)C=C2)C2=CC=CC=C12
1
–
1
of
1
results
1
R&D Systems™ LY 310762 hydrochloride
CAS: 192927-92-7 Molecular Formula: C24H28ClFN2O2