accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOH

CAS RN 19327-38-9

IUPAC Name: 2-{2-[2-(octyloxy)ethoxy]ethoxy}ethan-1-ol
Synonyms: 2-{2-[2-(octyloxy)ethoxy]ethoxy}ethanol
Molecular Weight (g/mol): 262.39
Molecular Formula: C14H30O4
InChi Key: XIVLVYLYOMHUGB-UHFFFAOYSA-N
SMILES: CCCCCCCCOCCOCCOCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results