CAS RN 193356-17-1
IUPAC Name:
1-[2-(4-hydroxyphenoxy)ethyl]-4-[(4-methylphenyl)methyl]piperidin-4-ol hydrochloride
Synonyms:
1-[2-(4-hydroxyphenoxy)ethyl]-4-[(4-methylphenyl)methyl]piperidin-4-ol hydrochloride
Molecular Weight (g/mol):
377.91
Molecular Formula:
C21H28ClNO3
InChi Key:
WFFZHKKSIDENAJ-UHFFFAOYSA-N
SMILES:
Cl.CC1=CC=C(CC2(O)CCN(CCOC3=CC=C(O)C=C3)CC2)C=C1
1
–
1
of
1
results
1
R&D Systems™ Co 101244 hydrochloride
CAS: 193356-17-1 Molecular Formula: C21H28ClNO3