accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNNNHOF

CAS RN 193551-21-2

IUPAC Name: 4-[4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)-1H-imidazol-1-yl]cyclohexan-1-ol
Synonyms: 4-[4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)imidazol-1-yl]cyclohexan-1-ol
Molecular Weight (g/mol): 368.41
Molecular Formula: C20H21FN4O2
InChi Key: ZQUSFAUAYSEREK-UHFFFAOYSA-N
SMILES: COC1=NC=CC(=N1)C1=C(N=CN1C1CCC(O)CC1)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)