accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19361-62-7

IUPAC Name: 1-[(1,2,2-²H₃)ethenyl](²H₅)benzene
Synonyms: 1-[(1,2,2-²H₃)ethenyl](²H₅)benzene
Molecular Weight (g/mol): 112.2
Molecular Formula: C8H8
InChi Key: PPBRXRYQALVLMV-GDALLCCDSA-N
SMILES: [2H]C([2H])=C([2H])C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results