Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Styrene-d8, for NMR, 98+ atom% D, stabilized

Catalog No. AC175390050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Ampoule
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC175390050 5 g Glass Bottle
AC175390010 1 g Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC175390050 Supplier Thermo Scientific Chemicals Supplier No. 175390050
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19361-62-7 | C8H8 | 112.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19361-62-7
Molecular Formula C8H8
Molecular Weight (g/mol) 112.201
MDL Number MFCD00044231
InChI Key PPBRXRYQALVLMV-GDALLCCDSA-N
Synonym styrene-d8, 2h8 styrene, perdeuteriostyrene, benzene-d5, ethenyl-d3, 1,2,3,4,5-pentadeuterio-6-1,2,2-trideuterioethenyl benzene, benzene-1,2,3,4,5-d5, 6-ethenyl-1,2,2-d3, styrene d8, phenethylene-d8, phenylethene-d8, vinylbenzene-d8
PubChem CID 88025
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(1,2,2-trideuterioethenyl)benzene
SMILES C=CC1=CC=CC=C1

Specifications

Physical Form Liquid
Flash Point 32°C
Packaging Glass Bottle
Color Colorless
Refractive Index 1.5443
Quantity 5 g
Density 0.9700g/mL
Deuterium (D) 98.0% min.
Water 0.05% max. (Coulometric)
Boiling Point 50.0°C to 51.0°C (25.0mmHg)
Linear Formula C6D5CD=CD2
Formula Weight 112.22
Grade NMR
Chemical Name or Material Styrene-d8, 98+ atom% D
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 242-995-6
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.