Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Styrene-d{8}, 98% (Isotopic) stab. with 4-tert-butylcatechol

Catalog No. AA4366703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4366703 1 g
AA4366706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4366703 Supplier Thermo Scientific Chemicals Supplier No. 04366703
Add to Cart

CAS: 19361-62-7 | C8H8 | 112.201 g/mol

Styrene-d{8} is used in Organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19361-62-7
Molecular Formula C8H8
Molecular Weight (g/mol) 112.201
MDL Number MFCD00044231
InChI Key PPBRXRYQALVLMV-GDALLCCDSA-N
Synonym styrene-d8, 2h8 styrene, perdeuteriostyrene, benzene-d5, ethenyl-d3, 1,2,3,4,5-pentadeuterio-6-1,2,2-trideuterioethenyl benzene, benzene-1,2,3,4,5-d5, 6-ethenyl-1,2,2-d3, styrene d8, phenethylene-d8, phenylethene-d8, vinylbenzene-d8
PubChem CID 88025
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(1,2,2-trideuterioethenyl)benzene
SMILES C=CC1=CC=CC=C1

Specifications

Physical Form Liquid
Sensitivity Air
Color Colorless
Quantity 1 g
Density 0.979 g/mL
Flash Point 31°C (88°F)
Refractive Index 1.5445
Packaging Ampule
Boiling Point 50°C to 51°C (25mmHg)
Linear Formula C6D5CD=CD2
UN Number UN2055
Beilstein 2327045
Solubility Information It is soluble in water 0.24g/L (20°C).
Formula Weight 112.2
Percent Purity 98%
Chemical Name or Material Styrene-d8, Stabilized with 4-tert-butylcatechol, Isotopic
Show More Show Less
Hazard Category H226-H304-H315-H319-H332-H335-H361d-H372
Hazard Statement GHS H Statement
H226-H332-H315-H319
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: STYRENE MONOMER, STABILIZED
EINECSNumber 242-995-6
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.