CAS RN 193611-67-5
IUPAC Name:
N-{3-[2-(dimethylamino)ethoxy]-4-methoxyphenyl}-2'-methyl-4'-(5-methyl-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-carboxamide hydrochloride
Synonyms:
N-{3-[2-(dimethylamino)ethoxy]-4-methoxyphenyl}-2'-methyl-4'-(5-methyl-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-carboxamide hydrochloride
Molecular Weight (g/mol):
523.03
Molecular Formula:
C28H31ClN4O4
InChi Key:
JPBMDMNORXKGHZ-UHFFFAOYSA-N
SMILES:
Cl.COC1=C(OCCN(C)C)C=C(NC(=O)C2=CC=C(C=C2)C2=C(C)C=C(C=C2)C2=NOC(C)=N2)C=C1
1
–
1
of
1
results
1
R&D Systems™ SB 216641 hydrochloride
CAS: 193611-67-5 Molecular Formula: C28H31ClN4O4