accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOHN

CAS RN 19395-39-2

IUPAC Name: 2-phenyl-2-(piperidin-2-yl)acetamide
Synonyms: 2-phenyl-2-(piperidin-2-yl)acetamide
Molecular Weight (g/mol): 218.3
Molecular Formula: C13H18N2O
InChi Key: LJLMNWPXAYKPGV-UHFFFAOYNA-N
SMILES: NC(=O)C(C1CCCCN1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)