accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1941-52-2

H3CSSH3COHHOOHHOOH

CAS RN 1941-52-2

IUPAC Name: 6,6-bis(ethylsulfanyl)hexane-1,2,3,4,5-pentol
Synonyms: 6,6-bis(ethylsulfanyl)hexane-1,2,3,4,5-pentol
Molecular Weight (g/mol): 286.4
Molecular Formula: C10H22O5S2
InChi Key: BTOYCPDACQXQRS-UHFFFAOYNA-N
SMILES: CCSC(SCC)C(O)C(O)C(O)C(O)CO
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)