Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucose Diethyl Mercaptal, TCI America™
SDP

Catalog No. G02591G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G02591G Supplier TCI America Supplier No. G02591G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1941-52-2
Molecular Formula C10H22O5S2
Molecular Weight (g/mol) 286.40
MDL Number MFCD00004706
InChI Key BTOYCPDACQXQRS-UHFFFAOYNA-N
Synonym d-glucose diethyl mercaptal, d-glucose diethyl dithioacetal, d-glucose diethylmercaptal, d-glucosediethylmercaptal, 2r,3r,4s,5r-6,6-bis ethylthio hexane-1,2,3,4,5-pentaol, 2r,3r,4s,5r-6,6-bis ethylsulfanyl hexane-1,2,3,4,5-pentol, d-glucose, diethyl dithioacetal, pubchem9903, opera_id_1736, glucose diethyl dithioacetal
PubChem CID 657375
IUPAC Name 6,6-bis(ethylsulfanyl)hexane-1,2,3,4,5-pentol
SMILES CCSC(SCC)C(O)C(O)C(O)C(O)CO

Specifications

Melting Point 128°C
Color White
Quantity 1 g
Formula Weight 286.40
Physical Form Crystalline Powder
Chemical Name or Material D-Glucose Diethyl Mercaptal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.