accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaNaH3CCH3H3CNOOOOOHOSOOCH3CH3OHNOOOOH3C

CAS RN 1945-77-3

IUPAC Name: tetrasodium 2-[({3-[3-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-6-hydroxy-2-methyl-5-(propan-2-yl)phenyl}methyl)(carboxylatomethyl)amino]acetate
Synonyms: tetrasodium 2-[({3-[3-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxo-2,1λ⁶-benzoxathiol-3-yl]-6-hydroxy-5-isopropyl-2-methylphenyl}methyl)(carboxylatomethyl)amino]acetate
Molecular Weight (g/mol): 844.75
Molecular Formula: C37H40N2Na4O13S
InChi Key: DIZZDZCUMBBRSG-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].CC(C)C1=CC(=C(C)C(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1C
Quantity 
  • (3)
  • (2)
  • (6)
Molecular Weight (g/mol) 
  • (2)
  • (8)
Color 
  • (3)
  • (3)
  • (1)
  • (2)
Grade 
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (1)
  • (3)
  • (3)
  • (4)
  • (3)
  • (2)
  • (6)
  • (2)
  • (8)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)