Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Methylthymol Blue sodium salt

Catalog No. AAL0938106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0938106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0938106 Supplier Thermo Scientific Chemicals Supplier No. L0938106
  • $146.20
    $108.65 / Each of 1

    Save $37.55

    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1945-77-3 | C37H44N2Na4O13S | 848.779 g/mol

Methylthymol Blue sodium salt is used as thymol-containing organic dye. It is used as an indicator for complexometry. It is used in determination of sulfate in rainwater. It combines with silica gel and aids in the separation and preconcentration of trace amounts of heavy metals from some salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylthymol Blue sodium salt is used as thymol-containing organic dye. It is used as an indicator for complexometry. It is used in determination of sulfate in rainwater. It combines with silica gel and aids in the separation and pr
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1945-77-3
Molecular Formula C37H44N2Na4O13S
Molecular Weight (g/mol) 848.779
MDL Number MFCD00148905
InChI Key LGVVZVZPOQJZKT-UHFFFAOYSA-N
Synonym Methylthymol Blue, water soluble
PubChem CID 131850582
IUPAC Name 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na]

Specifications

Solubility Information Soluble in water (460g/L) at 20°C.
Quantity 5 g
Formula Weight 844.76
Chemical Name or Material Methylthymol Blue sodium salt
EINECSNumber 217-743-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.