CAS RN 1949-89-9
MilliporeSigma™ 2-Deoxy-D-galactose, ≥99%, Calbiochem™,
CAS: 1949-89-9 Molecular Formula: C6H12O5 Molecular Weight (g/mol): 164.16 MDL Number: MFCD00014649 InChI Key: PMMURAAUARKVCB-ONLQHTGVNA-N Synonym: 2-deoxy-d-galactose,3r,4r,5r-3,4,5,6-tetrahydroxyhexanal,d-lyxo-hexose, 2-deoxy,unii-531k2iok5q,2-deoxy-d-lyxo-hexose,lyxo-hexose, 2-deoxy,ksc179c4l PubChem CID: 102191 ChEBI: CHEBI:27411 IUPAC Name: (4R,5R,6R)-6-(hydroxymethyl)oxane-2,4,5-triol SMILES: OC[C@H]1OC(O)C[C@@H](O)[C@H]1O
2-Deoxy-D-Galactose, MP Biomedicals™
CAS: 1949-89-9 Molecular Formula: C6H12O5 Molecular Weight (g/mol): 164.16 MDL Number: MFCD00014649 InChI Key: PMMURAAUARKVCB-ONLQHTGVNA-N Synonym: 2-deoxy-d-galactose,3r,4r,5r-3,4,5,6-tetrahydroxyhexanal,d-lyxo-hexose, 2-deoxy,unii-531k2iok5q,2-deoxy-d-lyxo-hexose,lyxo-hexose, 2-deoxy,ksc179c4l PubChem CID: 102191 ChEBI: CHEBI:27411 IUPAC Name: (4R,5R,6R)-6-(hydroxymethyl)oxane-2,4,5-triol SMILES: OC[C@H]1OC(O)C[C@@H](O)[C@H]1O
Thermo Scientific Chemicals 2-Deoxy-D-galactose, 99%
CAS: 1949-89-9 Molecular Formula: C6H12O5 Molecular Weight (g/mol): 164.16 MDL Number: MFCD00014649 InChI Key: PMMURAAUARKVCB-ONLQHTGVNA-N Synonym: 2-deoxy-d-galactose,3r,4r,5r-3,4,5,6-tetrahydroxyhexanal,d-lyxo-hexose, 2-deoxy,unii-531k2iok5q,2-deoxy-d-lyxo-hexose,lyxo-hexose, 2-deoxy,ksc179c4l PubChem CID: 102191 ChEBI: CHEBI:27411 IUPAC Name: (3R,4R,5R)-3,4,5,6-tetrahydroxyhexanal SMILES: OC[C@H]1OC(O)C[C@@H](O)[C@H]1O
2-deoxy-D-galactopyranose, MP Biomedicals™
CAS: 1949-89-9 Molecular Formula: C6H12O5 Molecular Weight (g/mol): 164.16 MDL Number: MFCD00014649 InChI Key: PMMURAAUARKVCB-ONLQHTGVNA-N Synonym: 2-deoxy-d-galactose,3r,4r,5r-3,4,5,6-tetrahydroxyhexanal,d-lyxo-hexose, 2-deoxy,unii-531k2iok5q,2-deoxy-d-lyxo-hexose,lyxo-hexose, 2-deoxy,ksc179c4l PubChem CID: 102191 ChEBI: CHEBI:27411 SMILES: OC[C@H]1OC(O)C[C@@H](O)[C@H]1O
2-Deoxy-D-Galactose, MP Biomedicals™
CAS: 1949-89-9 Molecular Formula: C6H12O5 Molecular Weight (g/mol): 164.16 MDL Number: MFCD00014649 InChI Key: PMMURAAUARKVCB-ONLQHTGVNA-N Synonym: 2-deoxy-d-galactose,3r,4r,5r-3,4,5,6-tetrahydroxyhexanal,d-lyxo-hexose, 2-deoxy,unii-531k2iok5q,2-deoxy-d-lyxo-hexose,lyxo-hexose, 2-deoxy,ksc179c4l PubChem CID: 102191 ChEBI: CHEBI:27411 IUPAC Name: (4R,5R,6R)-6-(hydroxymethyl)oxane-2,4,5-triol SMILES: OC[C@H]1OC(O)C[C@@H](O)[C@H]1O
Thermo Scientific Chemicals 2-Deoxy-D-galactose, 99%
CAS: 1949-89-9 Molecular Formula: C6H12O5 Molecular Weight (g/mol): 164.16 MDL Number: MFCD00014649 InChI Key: PMMURAAUARKVCB-ONLQHTGVNA-N Synonym: 2-deoxy-d-galactose,3r,4r,5r-3,4,5,6-tetrahydroxyhexanal,d-lyxo-hexose, 2-deoxy,unii-531k2iok5q,2-deoxy-d-lyxo-hexose,lyxo-hexose, 2-deoxy,ksc179c4l PubChem CID: 102191 ChEBI: CHEBI:27411 IUPAC Name: (3R,4R,5R)-3,4,5,6-tetrahydroxyhexanal SMILES: OC[C@H]1OC(O)C[C@@H](O)[C@H]1O