Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Deoxy-D-Galactose, MP Biomedicals™

Catalog No. MP021508005 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021508005 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021508005 Supplier MP Biomedicals, Inc Supplier No. 02150800.5
Add to Cart
Add to Cart

Inhibitor of fucosylation. 2-Deoxy-D-Galactose, MP Biomedicals is a hexose analog that can be incorporated into rat gangliosides GM2 and GD3 in positions normally occupied by galactose.

  • Hexose analog that can be incorporated into rat gangliosides GM2 and GD3 in positions normally occupied by galactose
  • Has been suggested to act as an inhibitor of fucosylation
  • Also used for competitive elution of Anadarin P lectin (a galactosyl-binding lectin from blood clam)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1949-89-9
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00014649
InChI Key PMMURAAUARKVCB-ONLQHTGVNA-N
Synonym 2-deoxy-d-galactose, 3r,4r,5r-3,4,5,6-tetrahydroxyhexanal, d-lyxo-hexose, 2-deoxy, unii-531k2iok5q, 2-deoxy-d-lyxo-hexose, lyxo-hexose, 2-deoxy, ksc179c4l
PubChem CID 102191
ChEBI CHEBI:27411
IUPAC Name (4R,5R,6R)-6-(hydroxymethyl)oxane-2,4,5-triol
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@H]1O

Specifications

Melting Point 109°C to 119°C
Quantity 500 mg
Optical Rotation 54.8°–66.3° (c=4, H2O)
Formula Weight 164.2
Physical Form Powder
Recommended Storage +4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.